Assay Details
| Gene Symbol: PABIR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614514.3 | XP_005614571.1 | 548 | 86 | ||
| XM_005614515.3 | XP_005614572.1 | 548 | 86 | |||
| XM_005614516.3 | XP_005614573.1 | 487 | 86 | |||
| XM_014729197.2 | XP_014584683.1 | 548 | 86 | |||
| XM_014729199.2 | XP_014584685.1 | 544 | 86 | |||
| GenBank mRNA | JL636786.1 | - | 4 - 5 | 340 | 86 | |
More Information
Set Membership: |
|

PABIR3
Genomic Map