Assay Details
| Gene Symbol: LOC100629745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362722.4 | XP_003362770.1 | 512 | 137 | ||
| XM_014730063.2 | XP_014585549.1 | 1652 | 137 | |||
| GenBank mRNA | JL639034.1 | - | 3 - 4 | 359 | 135 | |
More Information
Set Membership: |
|

LOC100629745
Genomic Map