Assay Details
| Gene Symbol: KLHL25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023650541.1 | XP_023506309.1 | 249 | 88 | ||
| XM_023650548.1 | XP_023506316.1 | 249 | 88 | |||
| XM_023650556.1 | XP_023506324.1 | 249 | 88 | |||
More Information
Set Membership: |
|

KLHL25
Genomic Map