Assay Details
| Gene Symbol: AHNAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023628409.1 | XP_023484177.1 | 6462 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AHNAK2| Gene Symbol: AHNAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023628409.1 | XP_023484177.1 | 6462 | 91 | ||