Assay Details
| Gene Symbol: NAA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492509.4 | XP_001492559.1 | 82 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001492509.4 transcripts.
Set Membership: |
|

NAA11
Genomic Map