Assay Details
| Gene Symbol: TSHZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647997.1 | XP_023503765.1 | 1170 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023647997.1 transcripts.
Set Membership: |
|

TSHZ1
Genomic Map