Assay Details
Gene Symbol: LOC100058633 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014739647.2 | XP_014595133.2 | 1604 | 95 | ||
XM_023640818.1 | XP_023496586.1 | 1640 | 95 | |||
XM_023640819.1 | XP_023496587.1 | 1625 | 95 | |||
XM_023640821.1 | XP_023496589.1 | 1614 | 95 | |||
XM_023640822.1 | XP_023496590.1 | 1599 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
