Assay Details
| Gene Symbol: LOC100058633 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014739647.2 | XP_014595133.2 | 479 | 103 | ||
| XM_023640818.1 | XP_023496586.1 | 479 | 103 | |||
| XM_023640819.1 | XP_023496587.1 | 464 | 103 | |||
| XM_023640821.1 | XP_023496589.1 | 453 | 103 | |||
| XM_023640822.1 | XP_023496590.1 | 438 | 103 | |||
More Information
Set Membership: |
|

LOC100058633
Genomic Map