Assay Details
Gene Symbol: LOC102150150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005609900.3 | XP_005609957.1 | 738 | 88 | ||
XM_005609902.3 | XP_005609959.1 | 738 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
