Assay Details
| Gene Symbol: KIRREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023640929.1 | XP_023496697.1 | 2221 | 83 | ||
| XM_023640930.1 | XP_023496698.1 | 2173 | 83 | |||
| XM_023640931.1 | XP_023496699.1 | 2125 | 83 | |||
| XM_023640932.1 | XP_023496700.1 | 2071 | 83 | |||
More Information
Set Membership: |
|

KIRREL1
Genomic Map