Assay Details
| Gene Symbol: NAT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023650540.1 | XP_023506308.1 | 303 | 94 | ||
| XM_023650542.1 | XP_023506310.1 | 200 | 94 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NAT14| Gene Symbol: NAT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023650540.1 | XP_023506308.1 | 303 | 94 | ||
| XM_023650542.1 | XP_023506310.1 | 200 | 94 | |||
Set Membership: |
|