Assay Details
| Gene Symbol: KCNJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498612.5 | XP_001498662.1 | 158 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001498612.5 transcripts.
Set Membership: |
|

KCNJ2
Genomic Map