Assay Details
Gene Symbol: COA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493268.4 | XP_001493318.1 | 273 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
