Assay Details
| Gene Symbol: PIGW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597546.3 | XP_005597603.1 | 1117 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005597546.3 transcripts.
Set Membership: |
|

PIGW
Genomic Map