Assay Details
Gene Symbol: MSX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502790.5 | XP_001502840.1 | 474 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
