Assay Details
Gene Symbol: GP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600339.3 | XP_005600396.2 | 2879 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005600339.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
