Assay Details
| Gene Symbol: CCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005600755.3 | XP_005600812.1 | 184 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005600755.3 transcripts.
Set Membership: |
|

CCR3
Genomic Map