Assay Details
Gene Symbol: LSM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499047.6 | XP_001499097.1 | 263 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001499047.6 transcripts.
Set Membership: |
![]() ![]() ![]() |
