Assay Details
Gene Symbol: LOC100066851 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636295.1 | XP_023492063.1 | 1247 | 65 | ||
XM_023636299.1 | XP_023492067.1 | 1244 | 65 | |||
XM_023636305.1 | XP_023492073.1 | 1258 | 65 | |||
XM_023636308.1 | XP_023492076.1 | 1261 | 65 | |||
XM_023636309.1 | XP_023492077.1 | 1261 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
