Assay Details
Gene Symbol: LOC100062243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503075.3 | XP_001503125.3 | 837 | 109 | ||
XM_023643143.1 | XP_023498911.1 | 723 | 109 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
