Assay Details
| Gene Symbol: ZNF248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646775.1 | XP_023502543.1 | 1005 | 85 | ||
| XM_023646779.1 | XP_023502547.1 | 1005 | 85 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023646775.1, XM_023646779.1 transcripts.
Set Membership: |
|

ZNF248
Genomic Map