Assay Details
Gene Symbol: LOC100062210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005602712.3 | XP_005602769.2 | 1081 | 63 | ||
XM_023647955.1 | XP_023503723.1 | 1078 | 63 | |||
XM_023647957.1 | XP_023503725.1 | 1081 | 63 | |||
XM_023647962.1 | XP_023503730.1 | 1081 | 63 | |||
XM_023647966.1 | XP_023503734.1 | 1081 | 63 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
