Assay Details
Gene Symbol: LOC102148936 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005602734.3 | XP_005602791.1 | 1258 | 97 | ||
XM_023648573.1 | XP_023504341.1 | 943 | 97 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
