Assay Details
| Gene Symbol: LOC102148936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005602734.3 | XP_005602791.1 | 1657 | 82 | ||
| XM_023648573.1 | XP_023504341.1 | 1342 | 82 | |||
More Information
Set Membership: |
|

LOC102148936
Genomic Map