Assay Details
Gene Symbol: RAMAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502380.6 | XP_001502430.1 | 108 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001502380.6 transcripts.
Set Membership: |
![]() ![]() |
