Assay Details
Gene Symbol: LOC100053576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003363593.4 | XP_003363641.2 | 1668 | 76 | ||
XM_014733927.2 | XP_014589413.1 | 1668 | 76 | |||
XM_023649788.1 | XP_023505556.1 | 1668 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
