Assay Details
Gene Symbol: LOC100053576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003363593.4 | XP_003363641.2 | 2208 | 86 | ||
XM_014733926.2 | XP_014589412.1 | 2198 | 86 | |||
XM_014733927.2 | XP_014589413.1 | 2130 | 86 | |||
XM_023649786.1 | XP_023505554.1 | 2121 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
