Assay Details
Gene Symbol: RNASE9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502624.4 | XP_001502674.2 | 287 | 85 | ||
XM_023621064.1 | XP_023476832.1 | 295 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001502624.4, XM_023621064.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
