Assay Details
Gene Symbol: ZNF146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005596190.2 | XP_005596247.1 | 142 | 98 | ||
XM_023651357.1 | XP_023507125.1 | 142 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005596190.2, XM_023651357.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
