Assay Details
Gene Symbol: ZNF146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005596190.2 | XP_005596247.1 | 373 | 76 | ||
XM_005596191.2 | XP_005596248.1 | 419 | 76 | |||
XM_005596194.2 | XP_005596251.1 | 331 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005596190.2, XM_005596191.2, XM_005596194.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
