Assay Details
| Gene Symbol: ZNF793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493662.6 | XP_001493712.3 | 292 | 84 | ||
| XM_005596212.3 | XP_005596269.1 | 291 | 84 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001493662.6, XM_005596212.3 transcripts.
Set Membership: |
|

ZNF793
Genomic Map