Assay Details
Gene Symbol: ZNF793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493662.6 | XP_001493712.3 | 562 | 99 | ||
XM_005596212.3 | XP_005596269.1 | 561 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001493662.6, XM_005596212.3 transcripts.
Set Membership: |
![]() ![]() |
