Assay Details
Gene Symbol: ZNF790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493784.5 | XP_001493834.2 | 847 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001493784.5 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
