Assay Details
| Gene Symbol: LOC100629711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023649829.1 | XP_023505597.1 | 180 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023649829.1 transcripts.
Set Membership: |
|

LOC100629711
Genomic Map