Assay Details
| Gene Symbol: LOC100054684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596530.3 | XP_005596587.2 | 1235 | 83 | ||
| XM_014730730.2 | XP_014586216.2 | 1206 | 83 | |||
| XM_023650430.1 | XP_023506198.1 | 1237 | 83 | |||
| XM_023650431.1 | XP_023506199.1 | 1213 | 83 | |||
| XM_023650432.1 | XP_023506200.1 | 948 | 83 | |||
More Information
Set Membership: |
|

LOC100054684
Genomic Map