Assay Details
| Gene Symbol: ZNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014731293.2 | XP_014586779.1 | 122 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014731293.2 transcripts.
Set Membership: |
|

ZNF135
Genomic Map