Assay Details
| Gene Symbol: ZNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596615.3 | XP_005596672.1 | 276 | 57 | ||
| XM_014731289.2 | XP_014586775.1 | 160 | 57 | |||
| XM_014731293.2 | XP_014586779.1 | 192 | 57 | |||
| XM_023650633.1 | XP_023506401.1 | 285 | 57 | |||
More Information
Set Membership: |
|

ZNF135
Genomic Map