Assay Details
Gene Symbol: C5H1orf162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610329.2 | XP_005610386.1 | 549 | 88 | ||
XM_005610330.3 | XP_005610387.1 | 504 | 88 | |||
XM_023641426.1 | XP_023497194.1 | 838 | 88 | |||
GenBank mRNA | JL624161.1 | - | 3 - 4 | 212 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
