Assay Details
Gene Symbol: KCNC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496153.4 | XP_001496203.1 | 2410 | 70 | ||
XM_005610345.3 | XP_005610402.1 | 2410 | 70 | |||
XM_005610347.2 | XP_005610404.1 | 2410 | 70 | |||
XM_023641442.1 | XP_023497210.1 | 2410 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
