Assay Details
| Gene Symbol: LOC100060051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736574.2 | XP_014592060.2 | 139 | 62 | ||
| XM_023652051.1 | XP_023507819.1 | 117 | 62 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014736574.2, XM_023652051.1 transcripts.
Set Membership: |
|

LOC100060051
Genomic Map