Assay Details
| Gene Symbol: MARCHF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023652264.1 | XP_023508032.1 | 280 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023652264.1 transcripts.
Set Membership: |
|

MARCHF10
Genomic Map