Assay Details
Gene Symbol: LOC100063851 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597306.3 | XP_005597363.3 | 5766 | 74 | ||
XM_005597307.3 | XP_005597364.3 | 5694 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
