Assay Details
Gene Symbol: C11H17orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023652890.1 | XP_023508658.1 | 2976 | 64 | ||
XM_023652893.1 | XP_023508661.1 | 1060 | 64 | |||
XM_023652894.1 | XP_023508662.1 | 1509 | 64 | |||
XM_023652895.1 | XP_023508663.1 | 1285 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
