Assay Details
Gene Symbol: TLCD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653186.1 | XP_023508954.1 | 736 | 93 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TLCD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653186.1 | XP_023508954.1 | 736 | 93 |