Assay Details
Gene Symbol: CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503419.4 | XP_001503469.2 | 253 | 99 | ||
XM_005597801.3 | XP_005597858.1 | 310 | 99 | |||
XM_023653535.1 | XP_023509303.1 | 249 | 99 | |||
XM_023653536.1 | XP_023509304.1 | 268 | 99 | |||
XM_023653537.1 | XP_023509305.1 | 144 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
