Assay Details
Gene Symbol: LOC100073135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653539.1 | XP_023509307.1 | 654 | 70 | ||
XM_023653540.1 | XP_023509308.1 | 608 | 70 | |||
XM_023653541.1 | XP_023509309.1 | 306 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
