Assay Details
| Gene Symbol: FAM111A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598161.3 | XP_005598218.1 | 571 | 85 | ||
| XM_023654100.1 | XP_023509868.1 | 570 | 85 | |||
| XM_023654101.1 | XP_023509869.1 | 571 | 85 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005598161.3, XM_023654100.1, XM_023654101.1 transcripts.
Set Membership: |
|

FAM111A
Genomic Map