Assay Details
| Gene Symbol: LOC100061154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598193.3 | XP_005598250.1 | 163 | 68 | ||
| XM_023654120.1 | XP_023509888.1 | 166 | 68 | |||
| XM_023654121.1 | XP_023509889.1 | 166 | 68 | |||
| GenBank mRNA | JL641342.1 | - | 6 - 7 | 890 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005598193.3, XM_023654120.1, XM_023654121.1 transcripts.
Set Membership: |
|

LOC100061154
Genomic Map