Assay Details
| Gene Symbol: LOC100061154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598193.3 | XP_005598250.1 | 325 | 109 | ||
| XM_023654120.1 | XP_023509888.1 | 328 | 109 | |||
| XM_023654121.1 | XP_023509889.1 | 328 | 109 | |||
| GenBank mRNA | JL641342.1 | - | 5 - 6 | 728 | 109 | |
More Information
Set Membership: |
|

LOC100061154
Genomic Map