Assay Details
| Gene Symbol: ZNF394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598598.3 | XP_005598655.2 | 662 | 73 | ||
| XM_023616803.1 | XP_023472571.1 | 1283 | 73 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF394| Gene Symbol: ZNF394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598598.3 | XP_005598655.2 | 662 | 73 | ||
| XM_023616803.1 | XP_023472571.1 | 1283 | 73 | |||
Set Membership: |
|